Learn More
Trajan Scientific and Medical™ Autosampler Syringes with Cone Tipped Needle for Agilent GCs
Gives better performance and superior quality. Trajan Scientific and Medical Autosampler Syringes with Cone Tipped Needle for Agilent GCs has the knowledge and expertise to manufacture an SGE syringe.
1465.00 SEK - 8660.00 SEK
Specifications
Needle Gauge | 23 G |
---|---|
Diameter (Metric) Outer Needle | 0.63 mm |
Needle Point Style | Cone |
Product Code | Brand | Volume (Metric) | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Product Type | Price | Quantity & Availability | ||||
15208443
|
Trajan Scientific and Medical™
000610 |
1 μL | NanoVolume Agilent Syringe |
1465.00 SEK
Each |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10514365
|
Trajan Scientific and Medical™
002820 |
10 μL | Removable Needle Agilent Syringe with GT Plunger |
8660.00 SEK
Pack of 10 |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Specifically designed for Agilent instruments
Specifications
23 G | |
0.63 mm | |
Cone |