Learn More
Trajan Scientific and Medical™ Autosampler Syringes with Cone Tipped Needle for Thermo Scientific GCs
Gives better performance and superior quality. Trajan Scientific and Medical Autosampler Syringes with Cone Tipped Needle for Thermo Scientific GCs has the knowledge and expertise to manufacture an SGE syringe.
Brand:  Trajan Scientific and Medical™ 0029931
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15218473
  • 1290.00 SEK / Each
Estimated Shipment: 05-06-2024
to see stock.
Description
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Thermo Scientific instruments
Specifications
Specifications
26 G
Removable Needle CTC/Thermo (Classic Button) Syringe
Cone
0.47 mm
10 μL