Learn More
Trajan Scientific and Medical™ Autosampler Syringes with Cone Tipped Needle for Thermo Scientific GCs
Gives better performance and superior quality. Trajan Scientific and Medical Autosampler Syringes with Cone Tipped Needle for Thermo Scientific GCs has the knowledge and expertise to manufacture an SGE syringe.
Brand:  Trajan Scientific and Medical™ 000790
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15218443
  • 1211.00 SEK / Each
Estimated Shipment: 04-08-2021
Description
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Thermo Scientific instruments
Specifications
Specifications
26 G
0.47 mm
NanoVolume CTC/Thermo Syringe
0.22 mm
2 µL
Cone