missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ AUTS2 Polyclonal Antibody
GREENER_CHOICE

Produktkod. 13280049
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
Förpackningsstorlek:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
13280049 100 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 13280049 Leverantör Invitrogen™ Leverantörsnummer PA522057

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, H1299, HepG2, Raji, NIH-3T3. Predicted reactivity: Mouse (100%), Pig (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

AUTS2 also named as FBRSL2 or autism susceptibility candidate 2 is a 1259 amino acid protein, which belongs to the AUTS2 family. AUTS2 is highly expressed in brain, kidney and skeletal muscle. AUTS2 exists as four isoforms produced by alternative splicing events and is encoded by a gene located on human chromosome 7. The molecular weight of AUTS2 isoform is 65-70 kDa in the brain. The functional role of AUTS2 is not well known, although some studies have identified a putative role in transcriptional regulation during neuronal development.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen AUTS2
Användningsområden Western Blot
Klassificering Polyclonal
Koncentration 1 mg/mL
Konjugera Unconjugated
Formulering PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gen Auts2
Genaccessionsnr. A0A087WPF7, Q8WXX7
Gene Alias 2700063G02Rik; A730011F23Rik; autism susceptibility candidate 2; autism susceptibility gene 2 protein; Autism susceptibility gene 2 protein homolog; autism-related protein 1; AUTS2; D830032G16Rik; FBRSL2; Kiaa0442; MRD26
Gensymboler Auts2
Värd art Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human AUTS2. The exact sequence is proprietary.
Reningsmetod Antigen affinity chromatography
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 26053, 319974
Målarter Human, Mouse, Zebrafish
Innehåll och lagring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.