Läs mer
Tocris Bioscience™ Avasimibe
Beskrivning
Avasimibe is an ACAT inhibitor (IC50 = 3.3 μM). Inhibits cholesterol esterification and reduces plasma cholesterol levels in rats. Also inhibits tumor growth and prolongs survival time in a melanoma mouse model and reduces tumor growth and metastasis in a pancreatic cancer mouse model.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 2,6-Diisopropylphenyl (2-(2,4,6-triisopropylphenyl)acetyl)sulfamate |
| CAS | 166518-60-1 |
| Kvantitet | 5 mg |
| Molekylformel | C29H43NO4S |
| Renhet | 98% |