Läs mer
MedChemExpress Aviptadil
Produktkod. 16454957
Encompass_Preferred
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
16454957 50 mg
1 options
Produktkod. 16454957 Leverantör MedChemExpress Leverantörsnummer HYP0012
MedChemExpress Aviptadil is an analog vasoactive intestinal polypeptide (VIP) with potent vasodilatory effects. Aviptadil induces pulmonary vasodilation and inhibits vascular SMCs proliferation, platelet aggregation. Aviptadil can be used for the research of pulmonary fibrosis, pulmonary arterial hypertension (PAH) and SARS-CoV-2 caused respiratory failure, et al.
Specifikationer
Renhet 99.81%
CAS 40077-57-4
Innehåll och lagring Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
För användning med (applikation) Cancer-programmed cell death
Molekylvikt (g/mol) 3325.8 g/mol
Produkttyp Peptides
Kvantitet 50 mg
Frakt skick Room temperature
Löslighet H2O : 33.33 mg/mL (10.02 mM∣Need ultrasonic)
Synonym Vasoactive Intestinal Peptide (human, rat, mouse, rabbit, canine, porcine)