Läs mer
MedChemExpress Aviptadil
Produktkod. 16461768
Encompass_Preferred
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
16461768 5 mg
1 options
Produktkod. 16461768 Leverantör MedChemExpress Leverantörsnummer HYP0012
MedChemExpress Aviptadil is an analog vasoactive intestinal polypeptide (VIP) with potent vasodilatory effects. Aviptadil induces pulmonary vasodilation and inhibits vascular SMCs proliferation, platelet aggregation. Aviptadil can be used for the research of pulmonary fibrosis, pulmonary arterial hypertension (PAH) and SARS-CoV-2 caused respiratory failure, et al.
Specifikationer
Renhet 99.81%
CAS 40077-57-4
Innehåll och lagring Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
För användning med (applikation) Cancer-programmed cell death
Molekylvikt (g/mol) 3325.8 g/mol
Produkttyp Peptides
Kvantitet 5 mg
Frakt skick Room temperature
Löslighet H2O : 33.33 mg/mL (10.02 mM∣Need ultrasonic)
Synonym Vasoactive Intestinal Peptide (human, rat, mouse, rabbit, canine, porcine)