Läs mer
Avizafone Dihydrobromide, TRC
Produktkod. 30345292
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30345292 50 mg
1 options
Produktkod. 30345292 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA79468550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Avizafone Dihydrobromide
CAS 60067-15-4
Molekylformel C22H29Br2ClN4O3
Kvantitet 50 mg
Synonym L-Lysyl-N-(2-benzoyl-4-chlorophenyl)-N-methyl-glycinamide Dihydrobromide, Prodiazepam Dihydrobromide, Ro 03-7355/000 Dihydrobromide
InChI formel InChI=1S/C22H27ClN4O3.2BrH/c1-27(20(28)14-26-22(30)18(25)9-5-6-12-24)19-11-10-16(23)13-17(19)21(29)15-7-3-2-4-8-15;;/h2-4,7-8,10-11,13,18H,5-6,9,12,14,24-25H2,1H3,(H,26,30);2*1H/t18-;;/m0../s1
LEDER ClC1=CC(C(C2=CC=CC=C2)=O)=C(N(C(CNC([C@@H](N)CCCCN)=O)=O)C)C=C1.Br.Br
IUPAC-namn (S)-2,6-diamino-N-(2-((2-benzoyl-4-chlorophenyl)(methyl)amino)-2-oxoethyl)hexanamide
Molekylvikt (g/mol) 592.76
Formel vikt 590.03