Läs mer
Avridine, TRC
Produktkod. 30346081
Change view
Kvantitet:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30346081 25 mg
30337666 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30346081 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA79469525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Avridine
CAS 35607-20-6
Molekylformel C43 H90 N2 O2
Kvantitet 25 mg
Synonym 2,2'-[[3-(Dioctadecylamino)propyl]imino]bisethanol, CP 20961, CP-20961, N,N-Dioctadecyl-N',N'-bis(2-hydroxyethyl)-1,3-diaminopropane, N,N-Dioctadecyl-N',N'-bis(2-hydroxyethyl)propanediamine
InChI formel InChI=1S/C43H90N2O2/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-36-44(38-35-39-45(40-42-46)41-43-47)37-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h46-47H,3-43H2,1-2H3
LEDER CCCCCCCCCCCCCCCCCCN(CCCCCCCCCCCCCCCCCC)CCCN(CCO)CCO
IUPAC-namn 2-[3-(dioctadecylamino)propyl-(2-hydroxyethyl)amino]ethanol
Molekylvikt (g/mol) 667.187
Formel vikt 666.7