missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AWAT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25119-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes an enzyme belonging to the diacylglycerol acyltransferase family. This enzyme produces wax esters by the esterification of long chain (or wax) alcohols with acyl-CoA-derived fatty acids. It functions in lipid metabolism in the skin, mostly in undifferentiated peripheral sebocytes. This enzyme may also have acyl-CoA:retinol acyltransferase activities, where it catalyzes the synthesis of diacylglycerols and retinyl esters.Specifications
AWAT2 | |
Polyclonal | |
Unconjugated | |
AWAT2 | |
AWAT2, DC4, DGAT2L4, hDC4, hWS, MFAT, Wax synthase, WS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
158835, 245532, 302425 | |
-20°C | |
Liquid |
Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6E1M8, Q6E213 | |
Awat2 | |
AWAT2 Fusion Protein Ag18936 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |