missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AWAT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | AWAT2 |
---|---|
Concentration | 0.21 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16823665
|
Proteintech
25119-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16813665
|
Proteintech
25119-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes an enzyme belonging to the diacylglycerol acyltransferase family. This enzyme produces wax esters by the esterification of long chain (or wax) alcohols with acyl-CoA-derived fatty acids. It functions in lipid metabolism in the skin, mostly in undifferentiated peripheral sebocytes. This enzyme may also have acyl-CoA:retinol acyltransferase activities, where it catalyzes the synthesis of diacylglycerols and retinyl esters.Specifications
AWAT2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
Q6E1M8, Q6E213 | |
158835, 245532, 302425 | |
AWAT2 Fusion Protein Ag18936 | |
Primary | |
-20°C | |
AWAT2 |
0.21 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
AWAT2, DC4, DGAT2L4, hDC4, hWS, MFAT, Wax synthase, WS | |
Awat2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |