Läs mer
Axygen™ 384-brunnars skirtade PCR-mikroplattor
Handla allt Axygen Scientific Produkter
Färg:
Blue
Clear
Green
Klart
Orange
Red
Violet
Vitt
Yellow
Volym (metrisk) Tja:
40 μL
50 μL
50μ L
55μ L
Inkluderar:
Double notch
Dubbla hack
Enstaka hack
Single notch
Förpackningsstorlek:
Paket med 10
Paket med 100
Paket med 20
Paket med 50
Beskrivning
Sealing Options: Axymats, heat sealing, adhesive films and foils.
- Available in 50μL and 55μL
- Single and double notch corner
Skirted (50μL)
- Application/Compatibility: standard thermal cycling
- Robotic-friendly full skirt design
- Packaging Description: 10 plates/Pk., 5 packs/Cs.
- Application/Compatibility: standard thermal cycling, fully ABI compatible for qPCR and sequencing
- Robotic-friendly full skirt design
- Packaging Description: 10 plates/Pk., 5 packs/Cs.
- Robotic-friendly rigid plate with full skirt
- Application/Compatibility: standard thermal cycling, ABI sequencing, automation applications
- Packaging Description: 10 plates/Pk., 2 packs/sleeve, 5 sleeves/Cs.
- Application/Compatibility: standard thermal cycling, ABI sequencing, qPCR
- Raised wells allow 55μL volume
- Packaging Description: 10 plates/Pk., 5 packs/Cs.
Colors available by special order.
Specifikationer
Specifikationer
| Streckkod | No Barcode |
| Certifikat och efterlevnad | RNase, DNase-free |
| Formatera | Standard |
| Array | 16 x 24 |
| Färg | Red |
| Kjol stil | Full |
| Material | Polypropylene |
| Volym (metrisk) Tja | 40 μL |
| Antal Wells | 384 |
| Volym (metrisk) Fungerar | 40 μL |
| Visa mer |