Learn More
Axygen™ Clear Hanging Format Automation Pipet Tips for Hamilton Microlab™ Star Line and Nimbus™, 5 Pack
Compatible with Hamilton Microlab™ Star Line and Nimbus™ automated liquid handling workstations
Brand:  Axygen™ HT-50-C-HTR-S-5
Additional Details : Weight : 8.47000kg
Product Code. 15551403
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Axygen offers an alternative choice for your Hamilton™ STARline system.
These tips are:
  • Manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality
  • Available in non-sterile, sterile, non-filtered, and filtered
  • Available as Clear or Conductive black polypropylene tips
  • Available in 96 well hanging tip rack, nested tip rack, and individual deck-seating rack format
  • Designed for Hamilton™ MICROLAB™ STAR, STARlet, STARplus and Nimbus™ workstations
Specifications
Specifications
50 μL
Sterile
Not Autoclavable
Non-pyrogenic, used for stringent specifications under strict process controls
Automation
Hamilton MICROLAB Nimbus
Non-Filtered
Clear
DNase, RNase and Pyrogen free
Hanging Tip Rack
5 Racks/Pk, 12 Pks/Cs.