Learn More
Axygen™ LightCycler 480 Compatible PCR Plates, 96-well
Product Code. 12739628 Shop All Axygen Scientific Products
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 50
Product Code. Color Unit Size
12739628 Clear Pack of 50
1 options
Product Code. 12739628 Supplier Axygen™ Supplier No. PCR96LC480CNF
  • ONLINE EXCLUSIVE PRICE
    3147.21 SEK
    3012.55 SEK / Paket med 50
    Save 134.66 SEK
    4% Av
Beräknad leverans: 03-03-2027
Add to basket
Add to basket
Bulk Quote Request
96-well plate is designed to fit the Roche LightCycler™ 480
  • Certified RNase- and DNase-free
  • Nonsterile
  • Large print alphanumeric grid for ease of well identification
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Certifications/Compliance RNase, DNase-free
Format Standard
Array 8 x 12
Color Clear
Product Type PCR Microplate
For Use With (Equipment) For Roche light cycler 480
No. of Wells 96
Sterility Non-sterile
Well Shape Round
For Use With (Application) RT-PCR
Includes Printed alphanumeric grid
No. per Case 50/Cs.
No. per Pack 10/Pk.