Läs mer
Axygen™ Axypet™ 12-Channel Digital Pipettor, Calibration 3x4 ALT
Handla allt Axygen Scientific Produkter
Volym (metrisk):
0,5 till 10 μl
20 till 200 μl
5 till 50 μl
50 till 300 μl
Förpackningsstorlek:
Varje
Beskrivning
- ISO 17025 Pre-calibrated
- Thumb-friendly soft spring pipetting system reduces fatigue
- Unique side EZIject button option reduces thumb movement
- Finger-hook provides for a more relaxed grip
- Large ergonomic push button is easy on the thumb during contact
- Locking feature prevents accidental volume changes during pipetting procedures
- Push button and ejector button are ergonomically located
- Curved ejector bar allows tips to be ejected in steps, reducing the mount of force required for ejection
- Tip manifold rotates 360°
- Each channel of the pipettor has an individual, precision piston assembly to ensure accuracy and reproducibility from one pipetting series to the next
- Shafts withdraw when pressed against a row of tips
- Each shaft moves independently, minimizing jamming of tips
- Micrometer is continuously adjustable for selection of whole or fractional volumes
Specifikationer
Specifikationer
| Certifikat och efterlevnad | ISO17025 |
| Antal kanaler | 12 |
| Produktlinje | Axypet |
| Typ | Multi-channel Pipettor |
| Volym (metrisk) | 0,5 till 10 μl |
| Kvantitet | 1/Pk., 1/Cs |