Läs mer
Axygen™ Axypet™ 12-Channel Digital Pipettor, Calibration 3x4 ALT
Produktkod. 17253614 Handla allt Axygen Scientific Produkter
Volym (metrisk):
0,5 till 10 μl
20 till 200 μl
5 till 50 μl
50 till 300 μl
Förpackningsstorlek:
Varje
Produktkod. 17253614
Brand: Axygen™ AP12300ALT
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
High levels of accuracy and precision with design innovations to make pipetting more comfortable during prolonged periods of time.
  • ISO 17025 Pre-calibrated
  • Thumb-friendly soft spring pipetting system reduces fatigue
  • Unique side EZIject button option reduces thumb movement
  • Finger-hook provides for a more relaxed grip
  • Large ergonomic push button is easy on the thumb during contact
  • Locking feature prevents accidental volume changes during pipetting procedures
  • Push button and ejector button are ergonomically located
  • Curved ejector bar allows tips to be ejected in steps, reducing the mount of force required for ejection
  • Tip manifold rotates 360°
  • Each channel of the pipettor has an individual, precision piston assembly to ensure accuracy and reproducibility from one pipetting series to the next
  • Shafts withdraw when pressed against a row of tips
  • Each shaft moves independently, minimizing jamming of tips
  • Micrometer is continuously adjustable for selection of whole or fractional volumes
TRUSTED_SUSTAINABILITY
Specifikationer
Certifikat och efterlevnad ISO17025
Antal kanaler 12
Produktlinje Axypet
Typ Multi-channel Pipettor
Volym (metrisk) 50 till 300 μl
Kvantitet 1/Pk., 1/Cs