Läs mer
Tocris Bioscience™ AZ 13705339
GREENER_CHOICE
Produktkod. 18788926
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18788926 10 mg
18766136 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18788926 Leverantör Tocris Bioscience™ Leverantörsnummer 6177/10
Highly potent PAK1 and PAK2 inhibitor
AZ 13705339 is a highly potent PAK1 and PAK2 inhibitor (IC50 values are 0.33 and 6 nM for PAK1 and PAK2, respectively). Displays >7500-fold selectivity for PAK1 over PAK4 (IC50 = 2.6 μM).
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-[[[2-[[3-(Ethylsulfonyl)-4-(4-methyl-1-piperazinyl)phenyl]amino]-5-fluoro-4-pyrimidinyl][5-(hydroxymethyl)-2-methylphenyl]amino]methyl]benzonitrile
CAS 2016806-57-6
Kvantitet 10 mg
Molekylformel C33H36FN7O3S
Formel vikt 629.75
Renhet 98%