Läs mer
Tocris Bioscience™ AZ 191
Potent DYRK1B inhibitor
Brand: Tocris Bioscience™ 5232/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
AZ 191 is a potent DYRK1B inhibitor (IC50 values are 17, 88 and 1890 nM for DYRK1B, DYRK1A and DYRK2 respectively). Inhibits DYRK1B-induced Gi phase cell cycle arrest in Panc-1 cells in vitro.Specifikationer
| AZ 191 | |
| N-[2-Methoxy-4-(4-methyl-1-piperazinyl)phenyl]-4-(1-methyl-1H-pyrrolo[2,3-c]pyridin-3-yl)-2-pyrimidinamine | |
| Soluble to 100mM in DMSO and to 20mM in ethanol | |
| Bioactive Small Molecule |
| 1594092-37-1 | |
| 10 mg | |
| Solid |
Produkttitel
Research Use Only. Not For Human or Veterinary Use