Läs mer
Tocris Bioscience™ AZ 6102
Beskrivning
AZ 6102 is a potent TNKS2 and TNKS1 inhibitor (IC50 values are 1 and 3 nM, respectively). Exhibits >100-fold selectivity for TNKS1/2 over PARP-1, PARP-2 and PARP-6. Inhibits Wnt signaling in DLD-1 cells (IC50 = 5 nM). Exhibits moderate oral bioavailability.
Specifikationer
Specifikationer
| Kemiskt namn eller material | rel-2-[4-[6-[(3R,5S)-3,5-Dimethyl-1-piperazinyl]-4-methyl-3-pyridinyl]phenyl]-3,7-dihydro-7-methyl-4H-pyrrolo[2,3-d]pyrimidin-4-one |
| CAS | 1645286-75-4 |
| Kvantitet | 50 mg |
| Mål | Tankyrase Inhibitors |
| Molekylformel | C25H28N6O |
| Formel vikt | 428.53 |
| Renhet | >98% |