Läs mer
Tocris Bioscience™ AZ 6102
Potent TNKS1/2 inhibitor
Brand: Tocris Bioscience™ 5775/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
AZ 6102 is a potent TNKS2 and TNKS1 inhibitor (IC50 values are 1 and 3 nM, respectively). Exhibits >100-fold selectivity for TNKS1/2 over PARP-1, PARP-2 and PARP-6. Inhibits Wnt signaling in DLD-1 cells (IC50 = 5 nM). Exhibits moderate oral bioavailability.Specifikationer
| rel-2-[4-[6-[(3R,5S)-3,5-Dimethyl-1-piperazinyl]-4-methyl-3-pyridinyl]phenyl]-3,7-dihydro-7-methyl-4H-pyrrolo[2,3-d]pyrimidin-4-one | |
| 1645286-75-4 | |
| 10 mg |