Läs mer
Tocris Bioscience™ AZ 628
GREENER_CHOICE
Produktkod. 30001607
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30001607 50 mg
30001584 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30001607 Leverantör Tocris Bioscience™ Leverantörsnummer 4836/50
Potent Raf kinase inhibitor
Potent, ATP-competitive inhibitor of Raf kinases (IC50 values are 29, 34 and 105 nM for c-Raf1, B-RafV600E and wild-type B-Raf, respectively). Displays selectivity for Raf kinases over a panel of 150 other kinases; inhibits activation of tyrosine protein kinases such as VEGFR2, Lyn, Flt1 and Fms. Also inhibits growth, and induces cell cycle arrest and apoptosis in colon and melanoma cell lines with the B-RafV600E mutation.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 878739-06-1
Molekylformel C27H25N5O2
Specifikationer
Kemiskt namn eller material AZ 628
Kvantitet 50 mg
Formel vikt 451.52
Renhet >98%
Molekylvikt (g/mol) 451.53