missing translation for 'onlineSavingsMsg'
Learn More

Azadibenzocyclooctyne-PEG4-NHS ester, Thermo Scientific Chemicals

Catalog Number 15496209
Click to view available options
Quantity:
10 mg
25 mg
100 mg
This item is not returnable. View return policy
MFCD22380747
This item is not returnable. View return policy
MFCD22380747

Azadibenzocyclooctyne-PEG4-NHS ester is applied for protein-peptide conjugation, peptide-small molecule conjugation, 18F radiolabeling, protein-oligonucleotide conjugation and surface modification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azadibenzocyclooctyne-PEG4-NHS ester is applied for protein-peptide conjugation, peptide-small molecule conjugation, 18F radiolabeling, protein-oligonucleotide conjugation and surface modification.

Solubility
Soluble in DCM, DMF, DMSO and THF.

Notes
Moisture sensitive. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY
Color Orange to Brown
Quantity 10 mg
Sensitivity Moisture sensitive
Solubility Information Soluble in DCM,DMF,DMSO and THF.
Formula Weight 649.69
Physical Form Oil
Chemical Name or Material Azadibenzocyclooctyne-PEG4-NHS ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'