Läs mer
Tocris Bioscience™ AZD 1480
Potent and selective JAK2 inhibitor; antiangiogenic
Brand: Tocris Bioscience™ 5617/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
AZD 1480 is a potent and selective JAK2 inhibitor (IC50 < 3 nM). Exhibits >50-fold selectivity for JAK2 over JAK3. Exhibits antitumor effects and inhibits formation of lung metastases in a mouse renal cancer model. Also inhibits STAT3 signaling, tumor angiogenesis and myeloid cell migration in vivo.Specifikationer
| 5-Chloro-N2-[(1S)-1-(5-fluoro-2-pyrimidinyl)ethyl]-N4-(5-methyl-1H-pyrazol-3-yl)-2,4-pyrimidine-2,4-diamine | |
| 935666-88-9 | |
| 10 mg |