Läs mer
Tocris Bioscience™ AZD 2066
mGlu5 antagonist; orally bioavailable and brain penetrant
Brand: Tocris Bioscience™ 5614/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
AZD 2066 is a mGlu5 antagonist. Displays discriminative effects in rats. Orally bioavailable and brain penetrant.Specifikationer
| 4-[5-[(1R)-1-[5-(3-Chlorophenyl)-3-isoxazolyl]ethoxy]-4-methyl-4H-1,2,4-triazol-3-yl]pyridine | |
| 934282-55-0 | |
| 10 mg |