Läs mer
AZD-2098, TRC
Produktkod. 30179144
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179144 5 mg
30179097 1 mg
30179116 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179144 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA9654935MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 566203-88-1
Molekylformel C11H9Cl2N3O3S
Molekylvikt (g/mol) 334.18
LEDER COc1nccnc1NS(=O)(=O)c2cccc(Cl)c2Cl
Specifikationer
Kemiskt namn eller material AZD-2098
Kvantitet 5 mg
Synonym ,3-Dichloro-N-(3-methoxy-2-pyrazinyl)benzenesulfonamide, AZD 2098, AZD2098