Läs mer
AZD-5438, MedChemExpress
Produktkod. 16459476
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16459476 50 mg
16434236 100 mg
16412788 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16459476 Leverantör Medchem Express Leverantörsnummer HY10012
MedChemExpress AZD-5438 is a potent CDK1, CDK2, and CDK9 inhibitor, with IC50s of 16 nM, 6 nM, and 20 nM in cell-free assays, respectively. AZD-5438 shows less inhibition activity against GSK3β, CDK5 and CDK6 .
AZD-5438 is a potent CDK1, CDK2, and CDK9 inhibitor, with IC50s of 16 nM, 6 nM, and 20 nM in cell-free assays, respectively. AZD-5438 shows less inhibition activity against GSK3β, CDK5 and CDK6
Specifikationer
Kemiskt namn eller material AZD-5438
CAS 602306-29-6
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C18H21N5O2S
Kvantitet 50 mg
Löslighetsinformation DMSO : 100 mg/mL (269.21 mM; Need ultrasonic)
LEDER O=S(C1=CC=C(NC2=NC(C3=CN=C(N3C(C)C)C)=CC=N2)C=C1)(C)=O
Molekylvikt (g/mol) 371.46
Formel vikt 371.46
Procent renhet 99.55%
Kvalitet Research
Fysisk form Powder
För användning med (applikation) Cancer-Kinase/protease