Läs mer
AZD-5991, MedChemExpress
Produktkod. 16443408
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
Förpackningsstorlek:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16443408 10 mg 10mg
16429936 50 mg -
16416137 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16443408 Leverantör Medchem Express Leverantörsnummer HY101533
MedChemExpress AZD-5991 is a potent and selective Mcl-1 inhibitor with an IC50 of 0.7 nM in FRET assay and a Kd of 0.17 nM in surface plasmon resonance (SPR) assay.
AZD-5991 is a potent and selective Mcl-1 inhibitor with an IC50 of 0.7 nM in FRET assay and a Kd of 0.17 nM in surface plasmon resonance (SPR) assay.
Specifikationer
Kemiskt namn eller material AZD-5991
Fysisk form Powder
CAS 2143061-81-6
Färg White
Kvantitet 10 mg
Anteckningar om renhetsgrad Research
Molekylformel C35H34ClN5O3S2
För användning med (applikation) Cancer-programmed cell death
Löslighetsinformation DMSO : 25 mg/mL (37.19 mM; Need ultrasonic)
LEDER O=C(O)C1=C(CCCOC2=C(C=CC=C3)C3=CC4=C2)C5=C(N1C)C(C6=C(C)N(C)N=C6CSCC7=NN(C)C(CS4)=C7)=C(Cl)C=C5
Molekylvikt (g/mol) 672.26
Formel vikt 672.26
Procent renhet 98.36%