Läs mer
AZD-7648, MedChemExpress
Produktkod. 16497237
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
Förpackningsstorlek:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16497237 5 mg 5mg
16464528 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16497237 Leverantör Medchem Express Leverantörsnummer HY111783
MedChemExpress AZD-7648 is a potent, orally active, selective DNA-PK inhibitor with an IC50 of 0.6 nM. AZD-7648 induces apoptosis and shows antitumor activity.
AZD-7648 is a potent, orally active, selective DNA-PK inhibitor with an IC50 of 0.6 nM. AZD-7648 induces apoptosis and shows antitumor activity.
Specifikationer
Kemiskt namn eller material AZD-7648
CAS 2230820-11-6
Färg Gray
Anteckningar om renhetsgrad Research
Molekylformel C18H20N8O2
Kvantitet 5 mg
Löslighetsinformation DMSO : 5 mg/mL (13.14 mM; ultrasonic and warming and heat to 80°C)
LEDER CC1=CC2=NC=NN2C=C1NC3=NC=C(N(C)C(N4C5CCOCC5)=O)C4=N3
Molekylvikt (g/mol) 380.4
Formel vikt 380.4
Procent renhet 98.14%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease