Läs mer
AZD-8835, MedChemExpress
Produktkod. 16498127
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16498127 5 mg
16466016 100 mg
16455418 10 mg
16401607 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16498127 Leverantör Medchem Express Leverantörsnummer HY12869
MedChemExpress AZD8835 is a potent and selective inhibitor of PI3Kα and PI3Kδ with IC50s of 6.2 and 5.7 nM, respectively.
AZD8835 is a potent and selective inhibitor of PI3Kα and PI3Kδ with IC50s of 6.2 and 5.7 nM, respectively.
Specifikationer
Kemiskt namn eller material AZD-8835
Fysisk form Solid
CAS 1620576-64-8
Färg Yellow
Kvantitet 5 mg
Anteckningar om renhetsgrad Research
Molekylformel C22H31N9O3
För användning med (applikation) Cancer-Kinase/protease
Löslighetsinformation DMSO : 20 mg/mL (42.59 mM; Need ultrasonic)
LEDER O=C(N1CCC(C2=NN(CC)C(C3=NC(C4=NN=C(C(C)(C)C)O4)=C(N)N=C3)=N2)CC1)CCO
Molekylvikt (g/mol) 469.54
Formel vikt 469.54
Procent renhet 98.52%