Läs mer
AZD4573, MedChemExpress
Produktkod. 16447277
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
100 mg
Förpackningsstorlek:
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16447277 5 mg -
16483126 25 mg -
16435416 100 mg -
16430867 50 mg 50mg
16404568 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16447277 Leverantör Medchem Express Leverantörsnummer HY112088
MedChemExpress AZD4573 is a potent and highly selective CDK9 inhibitor (IC50 of <4 nM) that enables transient target engagement for the treatment of hematologic malignancies.
AZD4573 is a potent and highly selective CDK9 inhibitor (IC50 of <4 nM) that enables transient target engagement for the treatment of hematologic malignancies.
Specifikationer
Kemiskt namn eller material AZD4573
CAS 2057509-72-3
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C22H28ClN5O2
Kvantitet 5 mg
Löslighetsinformation DMSO : 50 mg/mL (116.30 mM; Need ultrasonic)
LEDER ClC1=CN=C(NC([C@H]2CCC[C@@H](NC(C)=O)C2)=O)C=C1C3=C(CC(C)(C)C4)N4N=C3
Molekylvikt (g/mol) 429.94
Formel vikt 429.94
Procent renhet 98.03%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease