Learn More
STEMCELL Technologies AZD8055
Product Code. 17110162
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17110162 10 mg 10mg
17100162 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17110162 Supplier STEMCELL Technologies Supplier No. 73004
 Restricted Item
Beräknad leverans: 25-11-2026
mTOR pathway inhibitor; Inhibits mTOR
  • AZD8055 is a potent, selective ATP-competitive inhibitor of mammalian target of rapamycin (mTOR), with an IC50 value of 0.8 nM (Chresta et al.) It is very specific with ∼1,000-fold selectivity for mTOR over all PI3K isoforms and exhibits no activity against a panel of 260 kinases at concentrations up to 10 μM (Chresta et al.)
Specifications
CAS 1009298-09-2
Synonym CCG-168
Quantity 10 mg
Molecular Formula C25H31N5O4
Purity ≥98%