Learn More
Azilsartan Amide Medoxomil, TRC
Product Code. 30332156
Change view
Click to view available options
Kvantitet:
100 mg
Product Code. Kvantitet
30332156 100 mg
1 options
Product Code. 30332156 Supplier Toronto Research Chemicals Supplier No. TRCA926915100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material Azilsartan Amide Medoxomil
CAS 1696392-12-7
Molekylformel C29H25N3O7
Kvantitet 100 mg
Synonym 1-[[2'-(Aminocarbonyl)[1,1'-biphenyl]-4-yl]methyl]-2-ethoxy-1H-benzimidazole-7-carboxylic Acid (5-Methyl-2-oxo-1,3-dioxol-4-yl)methyl Ester
InChI formel InChI=1S/C29H25N3O7/c1-3-36-28-31-23-10-6-9-22(27(34)37-16-24-17(2)38-29(35)39-24)25(23)32(28)15-18-11-13-19(14-12-18)20-7-4-5-8-21(20)26(30)33/h4-14H,3,15-16H2,1-2H3,(H2,30,33)
LEDER CCOc1nc2cccc(C(=O)OCC3=C(C)OC(=O)O3)c2n1Cc4ccc(cc4)c5ccccc5C(=O)N
IUPAC-namn (5-methyl-2-oxo-1,3-dioxol-4-yl)methyl 3-[[4-(2-carbamoylphenyl)phenyl]methyl]-2-ethoxybenzimidazole-4-carboxylate
Molekylvikt (g/mol) 527.52
Formel vikt 527.1693