Läs mer
Azilsartan Hydroxy Acid, TRC
Produktkod. 17963545
Change view
Kvantitet:
10 mg
50 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17963545 50 mg
17961635 10 mg
17914795 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17963545 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA92693050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1397836-49-5
Molekylformel C24 H22 N4 O4
Molekylvikt (g/mol) 430.462
Synonym 2-Ethoxy-1-[[2'-[(hydroxyamino)iminomethyl][1,1'-biphenyl]-4-yl]methyl]-1H-benzimidazole-7-carboxylic Acid;
IUPAC-namn 2-ethoxy-3-[[4-[2-(N-hydroxycarbamimidoyl)phenyl]phenyl]methyl]benzimidazole-4-carboxylic acid
LEDER CCOc1nc2cccc(C(=O)O)c2n1Cc3ccc(cc3)c4ccccc4C(=N)NO
Specifikationer
Kemiskt namn eller material Azilsartan Hydroxy Acid
Analyt- eller komponentnamn Azilsartan Hydroxy Acid
Frakt skick Room Temperature
InChI formel InChI=1S/C24H22N4O4/c1-2-32-24-26-20-9-5-8-19(23(29)30)21(20)28(24)14-15-10-12-16(13-11-15)17-6-3-4-7-18(17)22(25)27-31/h3-13,31H,2,14H2,1H3,(H2,25,27)(H,29,30)
Lösningstyp Neat
Industry Type Forensics
Kvantitet 50 mg
Formel vikt 430.1641