Learn More
Azlon™ Multipurpose Stoppers
Made of PVC
Brand:  Azlon PWJ020
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10034234
  • 622.58 SEK / Each
Estimated Shipment
27-12-2017
Add to basket
Description
Description
  • For use with or without pumps PWJ014/PWJ016/PWJ018
  • Consists of three parts, one core and two collars
  • Can be used in container openings from 30mm to 70mm diameter
  • Has two apertures, one for inserting tubes and a smaller aperture which acts as an air vent to allow siphoning
Specifications
Specifications
2 Apertures
PWJ016
–25 to 70°C
12mm
Multipurpose Stopper
PVC (Polyvinyl chloride)
Videos
Documentation
Technical Documents