Learn More
Azlon™ Three Compartment Polypropylene Gallipot Tray
Suitable for sampling, short term storage, micro-analysis or any other applications.
Brand: Azlon GWY530
Additional Details : Weight : 0.03000kg
Description
- Three compartment tray with two integral gallipots
- Made from medical grade polypropylene
- Suitable for sampling, short term storage, micro-analysis or any other applications
- Non-sterile - can be autoclavable at 121°C
Specifications
130mm | |
28 mm | |
Suitable for sampling, short term storage, micro-analysis or any other applications |
Polypropylene | |
Yes |