missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Azurocidin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563191
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Immunogen sequence: FPRFVNVTVT PEDQCRPNNV CTGVLTRRGG ICNGDGGTPL VCEGLAHGVA SFSLGPCGRG PDFFT Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 48%.
Azurocidin, also called heparin-binding protein (HBP), is an inactive homolog of serine proteinases residing in granulocytes. Initially it was thought to have only activity against gram-negative bacteria. Later it was also found to act against gram-positive bacteria and C. albicans. Azurocidin is considered a family member of polymorphonuclear leukocytes- derived antimicrobial proteins like defensins, LL-37 and lysozyme. Azurocidin has been recognized as a player in the activation and modulation of the immune response and may act to alarm the immune system. The cationic domain at one side of the protein is essential for its antimicrobial activity. It is stored in azurophil granules as well as secretory granules and as a result partly released at early stage of extravasation. Azurocidin is involved in chemotaxis and activation of monocytes, cytokine release and phagocytosis leading to more efficient bacterial clearance. The primary targeted environment of azurocidin are cells in the bloodstream, that is the endothelial lining, and the extravascular surroundings. The interaction of azurocidin with leucocytes is mediated via beta 2-intergrins. High plasma levels of azurocidin might help with the identification of patients which are at risk in developing sepsis.
Specifikationer
| Azurocidin | |
| Polyclonal | |
| Unconjugated | |
| AZU1 | |
| AZAMP; AZU; AZU1; Azurocidin; azurocidin 1; CAP37; cationic antimicrobial protein 37; cationic antimicrobial protein CAP37; HBP; Heparin-binding protein; hHBP; HUMAZUR; NAZC; neutrophil azurocidin; pHBP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 566 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P20160 | |
| AZU1 | |
| Recombinant protein corresponding to Human Azurocidin. Recombinant protein control fragment (Product #RP-101488). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering