Läs mer
b-Benzyl L-Aspartic Acid N-carboxyanhydride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | beta-Benzyl L-Aspartic Acid N-carboxyanhydride |
| CAS | 13590-42-6 |
| Molekylformel | C12 H11 N O5 |
| Kvantitet | 25 g |
| Synonym | (4S)-2,5-Dioxo-4-oxazolidineacetic Acid Phenylmethyl Ester, L-2,5-Dioxo-4-oxazolidineacetic Acid Benzyl Ester, L-Aspartic acid β-benzyl ester N-carboxylic acid anhydride, β-Benzyl L-aspartate N-carboxyanhydride |
| InChI formel | InChI=1S/C12H11NO5/c14-10(6-9-11(15)18-12(16)13-9)17-7-8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H,13,16)/t9-/m0/s1 |
| LEDER | O=C(C[C@@H]1NC(=O)OC1=O)OCc2ccccc2 |
| IUPAC-namn | benzyl 2-[(4S)-2,5-dioxo-1,3-oxazolidin-4-yl]acetate |
| Molekylvikt (g/mol) | 249.22 |
| Formel vikt | 249.0637 |