Läs mer
b-Cyano-L-alanine, TRC
Produktkod. 30362225
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30362225 50 mg
1 options
Produktkod. 30362225 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC95510050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material beta-Cyano-L-alanine
CAS 6232-19-5
Molekylformel C4H6N2O2
Kvantitet 50 mg
Synonym 3-Cyano-L-alanine, (2S)-2-Amino-3-cyanopropanoic Acid, L-2-Amino-3-cyanopropanoic Acid, L-3-Cyanoalanine, L-β-Cyanoalanine β-Cyano-L-alanine
InChI formel InChI=1S/C4H6N2O2/c5-2-1-3(6)4(7)8/h3H,1,6H2,(H,7,8)/t3-/m0/s1
LEDER N[C@@H](CC#N)C(O)=O
IUPAC-namn (2S)-2-amino-3-cyanopropanoic acid
Molekylvikt (g/mol) 114.1
Formel vikt 114.0429