missing translation for 'onlineSavingsMsg'
Läs mer

b-Estradiol-6-one 6-(O-carboxymethyloxime), TRC

Produktkod. 30379746
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30379746 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30379746 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE88808025MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Beta-Estradiol-6-one 6-(O-carboxymethyloxime)
CAS 35048-47-6
Anteckningar om renhetsgrad HPLC
Molekylformel C20 H25 N O5
Kvantitet 25 mg
Synonym 2-[[[(17β)-3,17-Dihydroxyestra-1,3,5(10)-trien-6-ylidene]amino]oxy]acetic Acid, Estrane Acetic Acid Deriv., 17β-Estradiol 6-(O-carboxymethyl)oxime, 6-Oxo-17β-estradiol 6-[O-(carboxymethyl)oxime], 6-Oxoestradiol (carboxymethyl)oxime, Estradiol 6-(O-carboxymethyl)oxime, Estradiol 6-carboxymethyloxime, Estrone 6-(O-carboxymethyl)oxime
InChI formel InChI=1S/C20H25NO5/c1-20-7-6-13-12-3-2-11(22)8-15(12)17(21-26-10-19(24)25)9-14(13)16(20)4-5-18(20)23/h2-3,8,13-14,16,18,22-23H,4-7,9-10H2,1H3,(H,24,25)/b21-17-/t13-,14-,16+,18+,20+/m1/s1
LEDER C[C@]12CC[C@H]3[C@@H](C\C(=N\OCC(=O)O)\c4cc(O)ccc34)[C@@H]1CC[C@@H]2O
IUPAC-namn 2-[(Z)-[(8R,9S,13S,14S,17S)-3,17-dihydroxy-13-methyl-8,9,11,12,14,15,16,17-octahydro-7H-cyclopenta[a]phenanthren-6-ylidene]amino]oxyacetic acid
Molekylvikt (g/mol) 359.42
Formel vikt 359.1733
Procent renhet >95
Visa mer Visa mindre
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.