Läs mer
Tocris Bioscience™ B I09
IRE1 endonuclease inhibitor; cell permeable
Brand: Tocris Bioscience™ 6009/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
B I09 is an IRE-1 RNase inhibitor (IC50 = 1.23 μM). Inhibits IRE-1/XBP1 pathway. Cell permeable. Inhibits growth of human chronic lymphocytic leukemia (CLL) cells in vitro and promotes CLL regression in a mouse model.Specifikationer
| Beige | |
| 1607803-67-7 | |
| 303.31g/mol | |
| Store at -20°C | |
| 10 mg |
| Inhibitor | |
| C16H17NO5 | |
| Soluble to 50mM in DMSO | |
| Inhibits Rnase |
Produkttitel
For Research Use Only