Läs mer
(-)-B-Methoxydiisopinocampheylborane hydrate, TRC
Produktkod. 30469892
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30469892 5 g
1 options
Produktkod. 30469892 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2697405G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (-)-B-Methoxydiisopinocampheylborane hydrate
CAS 85134-98-1
Molekylformel C21H37BO . x(H 2O)
Kvantitet 5 g
Synonym (-)-B-Methoxydiisopinocampheylborane, (-)-Diisopinocampheylmethoxyborane, (-)-Ipc2B(OMe), (-)-Methoxydiisopinocampheylborane, Methoxybis((1R,2S,3R,5R)-2,6,6-trimethylbicyclo[3.1.1]heptan-3-yl)borane
InChI formel InChI=1S/C21H37BO/c1-12-16-8-14(20(16,3)4)10-18(12)22(23-7)19-11-15-9-17(13(19)2)21(15,5)6/h12-19H,8-11H2,1-7H3/t12-,13-,14+,15+,16-,17-,18-,19-/m1/s1
LEDER CC1(C)[C@@H]2C[C@H]1C[C@@H](B([C@@H]3C[C@@H]4C[C@@H](C4(C)C)[C@H]3C)OC)[C@@H]2C
IUPAC-namn methoxybis((1R,2S,3R,5R)-2,6,6-trimethylbicyclo[3.1.1]heptan-3-yl)borane
Molekylvikt (g/mol) 316.3