Läs mer
b-Phenylethyl b-D-Glucoside, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | beta-Phenylethyl beta-D-Glucoside |
| CAS | 18997-54-1 |
| Molekylformel | C14 H20 O6 |
| Kvantitet | 50 mg |
| Synonym | 2-Phenylethyl-β-D-glucopyranoside, 2-Phenylethyl O-β-D-Glucopyranoside, 2-Phenylethyl β-D-Glucopyranoside, 2-Phenylethyl β-D-Glucoside, Phenethyl β-D-Glucopyranoside, Phenethyl β-D-Glucoside, Phenylethyl 2-Glucoside, Phenylethyl β-D-Glucopyranoside, β-Phenylethanol Glucoside |
| InChI formel | InChI=1S/C14H20O6/c15-8-10-11(16)12(17)13(18)14(20-10)19-7-6-9-4-2-1-3-5-9/h1-5,10-18H,6-8H2/t10-,11-,12+,13-,14-/m1/s1 |
| LEDER | OC[C@H]1O[C@@H](OCCc2ccccc2)[C@H](O)[C@@H](O)[C@@H]1O |
| IUPAC-namn | (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(2-phenylethoxy)oxane-3,4,5-triol |
| Molekylvikt (g/mol) | 284.3 |
| Formel vikt | 284.126 |