Läs mer
b-Phenylethyl Furoate, TRC
Produktkod. 30425709
Change view
Kvantitet:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30425709 5 g
30418428 50 g
30424174 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30425709 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP4006135G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7149-32-8
Molekylformel C13 H12 O3
Molekylvikt (g/mol) 216.23
IUPAC-namn 2-phenylethyl furan-2-carboxylate
LEDER O=C(OCCc1ccccc1)c2occc2
Specifikationer
Kemiskt namn eller material b-Phenylethyl Furoate
Kvantitet 5 g
InChI formel InChI=1S/C13H12O3/c14-13(12-7-4-9-15-12)16-10-8-11-5-2-1-3-6-11/h1-7,9H,8,10H2
Formel vikt 216.0786