Läs mer
(+)-b-Pinene, TRC
Produktkod. 30418194
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30418194 500 mg
1 options
Produktkod. 30418194 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP466618500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (1R)-(+)-beta-Pinene
CAS 19902-08-0
Molekylformel C10 H16
Kvantitet 500 mg
Synonym (1R,5R)-(+)-2(10)-Pinene, (1R)-6,6-Dimethyl-2-methylenebicyclo[3.1.1]heptane, (+)-2(10)-Pinene, (+)-β-Pinene, (1R)-(+)-β-Pinene, (1R,5R)-6,6-dimethyl-2-methylenebicyclo[3.1.1]heptane, d-β-Pinene, (+)-beta-Pinene
InChI formel InChI=1S/C10H16/c1-7-4-5-8-6-9(7)10(8,2)3/h8-9H,1,4-6H2,2-3H3/t8-,9-/m1/s1
LEDER CC1(C)[C@@H]2CCC(=C)[C@H]1C2
IUPAC-namn (1R,5R)-6,6-dimethyl-4-methylidenebicyclo[3.1.1]heptane
Molekylvikt (g/mol) 136.23
Formel vikt 136.1252