Läs mer
BOLA B.Safe Air Valve
Beskrivning
- B.Safe Air Valve body made of PP with connection (thread UNF 1/4 28 G) suitable for B.Safe Caps
- Lid with aeration opening and Luerlock connector, integrated non-return valve with upstream porous PTFE membrane (0.001 mm) including clip-on service life indicator
- Packing Unit pcs. 50 and 100
- Service life months 6
- Porous PTFE Membrane for pressure compensation and protection from contamination, including clip-on service life indicator
Specifikationer
Specifikationer
| Längd (metrisk) | 0.001 mm |
| Inkluderar | 6 Service Life Months |
| Material | PTFE |
| Typ | Valve |
| Kvantitet | 100-pack |