missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ B2M (Human) Recombinant Protein
Human B2M full-length ORF ( AAH32589, 1 a.a. - 119 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000567-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MSRSVALAVLALLSLSGLEAIQRTPKIQVYSRHPAENGKSNFLNCYVSGFHPSDIEVDLLKNGERIEKVEHSDLSFSKDWSFYLLYYTEFTPTEKDEYACRVNHVTLSQPKIVKWDRDM
Specifications
AAH32589 | |
beta-2-microglobulin | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
567 | |
Wheat germ expression system | |
25 μg | |
B2M | |
GST |