missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B3GNT9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566526
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: VFLGMCLQRLR LTPEPHPAFR TFGIPQPSAA PHLSTFD Highest antigen sequence identity to the following orthologs - mouse 95%, rat 95%.
B3GNT9 Polyclonal antibody specifically detects B3GNT9 in Human samples. It is validated for Immunocytochemistry.Specifications
B3GNT9 | |
Polyclonal | |
Unconjugated | |
B3GNT9 | |
B3GNT9; beta-1,3-galactosyltransferase-related protein; beta-1,3-Gn-T9; beta-1,3-N-acetylglucosaminyltransferase 9; beta3Gn-T9; BGnT-9; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 9; UNQ1922/PRO4397 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84752 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UX72 | |
B3GNT9 | |
Recombinant Human B3GNT9 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only