missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-H3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14453-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
CD276 is a member of the B7 family of co-stimulatory molecules also known as B7-H3. CD276 is a type I transmembrane protein that induces the proliferation of CD4+ and CD8+ T cells, enhances the generation of cytotoxic T cells and selectively stimulates the production of interferon gamma. Expression of CD276 can be induced on dendritic cells and monocytes by inflammatory cytokines, and is also widely expressed in peripheral tissues including the heart, kidney, testis and colon. In humans, CD276 exists as two isoforms which result from gene duplication and differential splicing. CD276 is reported to have therapeutic potential for the regulation of cell-mediated immune responses to cancer, particularly in conjunction with anti-angiogenic therapy.Specifications
B7-H3 | |
Polyclonal | |
Unconjugated | |
CD276 | |
4Ig B7 H3, B7 homolog 3, B7-H3, CD276, CD276 antigen, CD276 molecule, Costimulatory molecule, UNQ309/PRO352 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
80381 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5ZPR3 | |
CD276 | |
B7-H3 Fusion Protein Ag6006 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |