Läs mer
B7-H4 Monoclonal Antibody (H74), Functional Grade, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 16-5949-050
Beskrivning
The H74 MAb was generated and reacts with human B7-S1, also known as B7-H4 and B7x. Cross reactivity of this antibody to other proteins has not been determined. B7S1 is a newly discovered member of the B7 family. It is speculated that the costimulatory regulation of T cells by B7S1 is influenced by the activation status of B cells. While it is reported that BTLA is a counter receptor for B7S1, further studies are needed to definitely determine the B7-H4 ligand. H74 stains human B7-H4 transfected cells and not peripheral blood cells. Exact expression pattern of B7-H4 has not been fully characterized.
B7-H4 Monoclonal antibody specifically detects B7-H4 in Human samples. It is validated for Control, Flow Cytometry, Functional Assay
Specifikationer
| B7-H4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.2 | |
| Q7Z7D3 | |
| Mouse | |
| 50 mg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry, Functional Assay | |
| H74 | |
| Functional Grade | |
| Vtcn1 | |
| B7 family member, H4; B7 homolog 4; B7 superfamily member 1; B7h.5; B7h4; B7-H4; B7S1; B7x; BC032925; EMBL:AAH95842.1}; FLJ22418; Immune costimulatory protein B7-H4; PRO1291; Protein B7S1; RGD1311204; RP11-229A19.4; T cell costimulatory molecule B7x; T-cell costimulatory molecule B7x; UNQ659/PRO1291; VCTN1; V-set domain containing T cell activation inhibitor 1; V-set domain containing T-cell activation inhibitor 1; V-set domain-containing T-cell activation inhibitor 1; vtcn1; vtcn1 {ECO:0000312 | |
| Affinity chromatography | |
| RUO | |
| 79679 | |
| 4° C | |
| Liquid |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.