Läs mer
Bacampicillin hydrochloride, MedChemExpress
Produktkod. 16464557
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
Förpackningsstorlek:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16464557 50 mg -
16458678 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16464557 Leverantör Medchem Express Leverantörsnummer HYB1149A
MedChemExpress Bacampicillin hydrochloride is a penicillin antibiotic, is a prodrug of ampicillin with improved oral bioavailability.
Bacampicillin hydrochloride is a penicillin antibiotic, is a prodrug of ampicillin with improved oral bioavailability.
Specifikationer
Kemiskt namn eller material Bacampicillin hydrochloride
Fysisk form Powder
CAS 37661-08-8
Färg White
Kvantitet 50 mg
Anteckningar om renhetsgrad Research
Molekylformel C21H28ClN3O7S
För användning med (applikation) COVID-19-immunoregulation
Löslighetsinformation DMSO : 125 mg/mL (249.01 mM; Need ultrasonic)
LEDER O=C([C@@H](C(C)(C)S[C@]1([H])[C@@H]2NC([C@H](N)C3=CC=CC=C3)=O)N1C2=O)OC(OC(OCC)=O)C.[H]Cl
Molekylvikt (g/mol) 501.98
Formel vikt 501.98
Procent renhet 98.03%